Assay Details
| Gene Symbol: LOC101102169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015198.4 | XP_004015247.1 | 5 - 6 | 584 | 137 | |
| XM_012190353.3 | XP_012045743.1 | 5 - 6 | 587 | 137 | ||
| XM_027977414.1 | XP_027833215.1 | 1 - 2 | 479 | 137 | ||
More Information
Set Membership: |
|

LOC101102169
Genomic Map