Assay Details
| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968956.1 | XP_027824757.1 | 3 - 4 | 1449 | 60 | |
| XM_027968967.1 | XP_027824768.1 | 3 - 4 | 743 | 60 | ||
| XM_027968975.1 | XP_027824776.1 | 3 - 4 | 972 | 60 | ||
More Information
Set Membership: |
|

B4GALT2
Genomic Map