Assay Details
| Gene Symbol: TSC22D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003260.4 | XP_004003309.3 | 1 - 2 | 2953 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSC22D2| Gene Symbol: TSC22D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003260.4 | XP_004003309.3 | 1 - 2 | 2953 | 80 | |
Set Membership: |
|