Assay Details
| Gene Symbol: IFI30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205150.1 | NP_001192079.1 | 6 - 7 | 669 | 106 | |
| GenBank mRNA | HM017967.1 | - | 6 - 7 | 669 | 106 | |
More Information
Set Membership: |
|

IFI30
Genomic Map