Assay Details
| Gene Symbol: OTUD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963349.1 | XP_027819150.1 | 7 - 8 | 2253 | 64 | |
| XM_027963350.1 | XP_027819151.1 | 7 - 8 | 2252 | 64 | ||
| XM_027963351.1 | XP_027819152.1 | 7 - 8 | 2251 | 64 | ||
| XM_027963352.1 | XP_027819153.1 | 8 - 9 | 2084 | 64 | ||
More Information
Set Membership: |
|

OTUD5
Genomic Map