Assay Details
| Gene Symbol: RWDD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011130.4 | XP_004011179.1 | 1 - 2 | 1306 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RWDD2A| Gene Symbol: RWDD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011130.4 | XP_004011179.1 | 1 - 2 | 1306 | 71 | |
Set Membership: |
|