Assay Details
| Gene Symbol: COLEC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097602.2 | XP_014953088.2 | 3 - 4 | 373 | 62 | |
| XM_027973084.1 | XP_027828885.1 | 3 - 4 | 759 | 62 | ||
More Information
Set Membership: |
|

COLEC10
Genomic Map