Assay Details
| Gene Symbol: RAB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097461.2 | XP_014952947.2 | 1 - 2 | 386 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB32| Gene Symbol: RAB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097461.2 | XP_014952947.2 | 1 - 2 | 386 | 54 | |
Set Membership: |
|