Assay Details
| Gene Symbol: TIPARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003222.4 | XP_004003271.1 | 3 - 4 | 1338 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TIPARP| Gene Symbol: TIPARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003222.4 | XP_004003271.1 | 3 - 4 | 1338 | 69 | |
Set Membership: |
|