Assay Details
| Gene Symbol: TRIM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008737.3 | XP_004008786.3 | 5 - 6 | 2508 | 68 | |
| XM_027970048.1 | XP_027825849.1 | 5 - 6 | 2543 | 68 | ||
More Information
Set Membership: |
|

TRIM41
Genomic Map