Assay Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956505.1 | XP_027812306.1 | 4 - 5 | 463 | 64 | |
| XM_027956506.1 | XP_027812307.1 | 4 - 5 | 508 | 64 | ||
| XM_027956507.1 | XP_027812308.1 | 3 - 4 | 652 | 64 | ||
| XM_027956508.1 | XP_027812309.1 | 2 - 3 | 394 | 64 | ||
More Information
Set Membership: |
|

HVCN1
Genomic Map