Assay Details
| Gene Symbol: BZW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004812.3 | XP_004004861.1 | 7 - 8 | 762 | 66 | |
| XM_027965075.1 | XP_027820876.1 | 4 - 5 | 394 | 66 | ||
More Information
Set Membership: |
|

BZW1
Genomic Map