Assay Details
| Gene Symbol: LOC101104137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965897.1 | XP_027821698.1 | 14 - 15 | 1787 | 99 | |
| Gene Symbol: LOC114113719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965896.1 | XP_027821697.1 | 5 - 6 | 422 | 100 | |

LOC101104137
Genomic Map