Assay Details
| Gene Symbol: CA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002442.4 | XP_004002491.2 | 10 - 11 | 2514 | 71 | |
| XM_012180403.3 | XP_012035793.2 | 11 - 12 | 2221 | 71 | ||
| XM_027974293.1 | XP_027830094.1 | 9 - 10 | 2411 | 71 | ||
More Information
Set Membership: |
|

CA14
Genomic Map