Assay Details
| Gene Symbol: LOC101116039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967507.1 | XP_027823308.1 | 3 - 4 | 1134 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101116039| Gene Symbol: LOC101116039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967507.1 | XP_027823308.1 | 3 - 4 | 1134 | 110 | |
Set Membership: |
|