Assay Details
| Gene Symbol: LYRM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012489.4 | XP_004012538.1 | 2 - 3 | 234 | 69 | |
| XM_012185383.3 | XP_012040773.1 | 3 - 4 | 1286 | 69 | ||
| XM_012185385.3 | XP_012040775.1 | 3 - 4 | 346 | 69 | ||
| XM_027975243.1 | XP_027831044.1 | 2 - 3 | 474 | 69 | ||
More Information
Set Membership: |
|

LYRM9
Genomic Map