Assay Details
| Gene Symbol: GID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974795.1 | XP_027830596.1 | 1 - 2 | 365 | 71 | |
| XM_027974796.1 | XP_027830597.1 | 1 - 2 | 401 | 71 | ||
More Information
Set Membership: |
|

GID4
Genomic Map