Assay Details
| Gene Symbol: CRISP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185120.1 | NP_001172049.1 | 4 - 5 | 291 | 59 | |
| XM_027958339.1 | XP_027814140.1 | 3 - 4 | 230 | 59 | ||
| GenBank mRNA | GU985270.1 | - | 4 - 5 | 291 | 59 | |
More Information
Set Membership: |
|

CRISP2
Genomic Map