Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199068.1 | NP_001185997.1 | 4 - 5 | 635 | 62 | |
| XM_027977371.1 | XP_027833172.1 | 3 - 4 | 415 | 62 | ||
| GenBank mRNA | HQ433587.1 | - | 4 - 5 | 635 | 62 | |
More Information
Set Membership: |
|

NCR1
Genomic Map