Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009401.2 | NP_001009401.1 | 1 - 2 | 150 | 105 | |
| GenBank mRNA | KC912524.1 | - | 1 - 2 | 107 | 105 | |
| S74436.1 | - | 1 - 2 | 124 | 105 | ||
| X78306.1 | - | 152 | 105 | |||
More Information
Set Membership: |
|

CXCL8
Genomic Map