Assay Details
| Gene Symbol: SLC25A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127278.1 | NP_001120750.1 | 2 - 3 | 605 | 110 | |
| GenBank mRNA | EU626207.1 | - | 2 - 3 | 605 | 110 | |
More Information
Set Membership: |
|

SLC25A4
Genomic Map