Assay Details
| Gene Symbol: MYLPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145183.1 | NP_001138655.1 | 3 - 4 | 231 | 61 | |
| XM_012103914.3 | XP_011959304.1 | 3 - 4 | 315 | 61 | ||
| GenBank mRNA | FJ546077.1 | - | 3 - 4 | 231 | 61 | |
More Information
Set Membership: |
|

MYLPF
Genomic Map