Assay Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162547.1 | NP_001156019.1 | 1 - 2 | 72 | 89 | |
| GenBank mRNA | FJ969399.1 | - | 1 - 2 | 72 | 89 | |
More Information
Set Membership: |
|

ARF5
Genomic Map