Assay Details
| Gene Symbol: GPR50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009726.1 | NP_001009726.1 | 1 - 2 | 191 | 63 | |
| GenBank mRNA | AF047829.1 | - | 1 - 2 | 191 | 63 | |
More Information
Set Membership: |
|

GPR50
Genomic Map