Assay Details
| Gene Symbol: UQCRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145187.2 | NP_001138659.1 | 3 - 4 | 311 | 92 | |
| GenBank mRNA | FJ546083.1 | - | 3 - 4 | 312 | 92 | |
More Information
Set Membership: |
|

UQCRH
Genomic Map