Assay Details
| Gene Symbol: RHOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163596.1 | NP_001157068.1 | 1 - 2 | 210 | 116 | |
| XM_027971436.1 | XP_027827237.1 | 1 - 2 | 309 | 116 | ||
| GenBank mRNA | FJ937965.1 | - | 1 - 2 | 210 | 116 | |
More Information
Set Membership: |
|

RHOV
Genomic Map