Assay Details
| Gene Symbol: CIDEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306115.1 | NP_001293044.1 | 5 - 6 | 848 | 108 | |
| XM_004018278.3 | XP_004018327.1 | 5 - 6 | 1745 | 108 | ||
| XM_012099534.3 | XP_011954924.1 | 5 - 6 | 1005 | 108 | ||
| XM_012099535.3 | XP_011954925.1 | 5 - 6 | 598 | 108 | ||
| XM_027957552.1 | XP_027813353.1 | 3 - 4 | 427 | 108 | ||
| GenBank mRNA | KM199684.1 | - | 5 - 6 | 848 | 108 | |
More Information
Set Membership: |
|

CIDEC
Genomic Map