Assay Details
| Gene Symbol: MBOAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306122.1 | NP_001293051.1 | 2 - 3 | 345 | 86 | |
| GenBank mRNA | JQ685711.1 | - | 2 - 3 | 345 | 86 | |
More Information
Set Membership: |
|

MBOAT4
Genomic Map