Assay Details
| Gene Symbol: IRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009740.1 | NP_001009740.1 | 7 - 8 | 741 | 61 | |
| XM_012105645.3 | XP_011961035.1 | 8 - 9 | 968 | 61 | ||
| XM_027962476.1 | XP_027818277.1 | 8 - 9 | 987 | 61 | ||
| XM_027962477.1 | XP_027818278.1 | 9 - 10 | 1227 | 61 | ||
| GenBank mRNA | AF228445.1 | - | 7 - 8 | 741 | 61 | |
More Information
Set Membership: |
|

IRF2
Genomic Map