Assay Details
| Gene Symbol: IL23A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185122.2 | NP_001172051.1 | 2 - 3 | 406 | 78 | |
| GenBank mRNA | FN822243.2 | - | 2 - 3 | 406 | 78 | |
| JN935767.1 | - | 2 - 3 | 268 | 78 | ||
More Information
Set Membership: |
|

IL23A
Genomic Map