Assay Details
| Gene Symbol: APOBEC3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093784.1 | NP_001087253.1 | 3 - 4 | 572 | 69 | |
| GenBank mRNA | DQ974645.1 | - | 3 - 4 | 572 | 69 | |
More Information
Set Membership: |
|

APOBEC3F
Genomic Map