Assay Details
| Gene Symbol: PAX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178052.1 | NP_001171523.1 | 3 - 4 | 447 | 62 | |
| XM_027960132.1 | XP_027815933.1 | 4 - 5 | 983 | 62 | ||
| XM_027960133.1 | XP_027815934.1 | 4 - 5 | 980 | 62 | ||
| XM_027960134.1 | XP_027815935.1 | 4 - 5 | 983 | 62 | ||
| XM_027960135.1 | XP_027815936.1 | 4 - 5 | 980 | 62 | ||
| XM_027960136.1 | XP_027815937.1 | 4 - 5 | 982 | 62 | ||
| XM_027960137.1 | XP_027815938.1 | 4 - 5 | 1784 | 62 | ||
| XM_027960138.1 | XP_027815939.1 | 5 - 6 | 716 | 62 | ||
| XM_027960139.1 | XP_027815940.1 | 4 - 5 | 981 | 62 | ||
| XM_027960140.1 | XP_027815941.1 | 4 - 5 | 978 | 62 | ||
| XM_027960141.1 | XP_027815942.1 | 4 - 5 | 650 | 62 | ||
| XM_027960142.1 | XP_027815943.1 | 5 - 6 | 4431 | 62 | ||
| XM_027960143.1 | XP_027815944.1 | 6 - 7 | 4361 | 62 | ||
| XM_027960144.1 | XP_027815945.1 | 5 - 6 | 4235 | 62 | ||
| XM_027960145.1 | XP_027815946.1 | 6 - 7 | 3428 | 62 | ||
| XM_027960146.1 | XP_027815947.1 | 6 - 7 | 3232 | 62 | ||
| XM_027960147.1 | XP_027815948.1 | 5 - 6 | 749 | 62 | ||
| GenBank mRNA | FN600706.1 | - | 3 - 4 | 447 | 62 | |
More Information
Set Membership: |
|

PAX2
Genomic Map