Assay Details
| Gene Symbol: IFNAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009342.1 | NP_001009342.1 | 1 - 2 | 139 | 73 | |
| GenBank mRNA | U65979.1 | - | 1 - 2 | 139 | 73 | |
More Information
Set Membership: |
|

IFNAR2
Genomic Map