Assay Details
| Gene Symbol: MAPK14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142894.1 | NP_001136366.1 | 2 - 3 | 253 | 77 | |
| XM_027958335.1 | XP_027814136.1 | 2 - 3 | 621 | 77 | ||
| XM_027958336.1 | XP_027814137.1 | 2 - 3 | 644 | 77 | ||
| XM_027958337.1 | XP_027814138.1 | 2 - 3 | 637 | 77 | ||
| GenBank mRNA | FJ236706.1 | - | 2 - 3 | 253 | 77 | |
More Information
Set Membership: |
|

MAPK14
Genomic Map