Assay Details
| Gene Symbol: FGF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291082.1 | NP_001278011.1 | 1 - 2 | 630 | 67 | |
| GenBank mRNA | JQ941957.1 | - | 1 - 2 | 630 | 67 | |
More Information
Set Membership: |
|

FGF5
Genomic Map