Assay Details
| Gene Symbol: RAB9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161870.1 | NP_001155342.1 | 1 | 518 | 70 | |
| XM_027962749.1 | XP_027818550.1 | 5 | 887 | 70 | ||
| XM_027962750.1 | XP_027818551.1 | 4 | 748 | 70 | ||
| XM_027962751.1 | XP_027818552.1 | 3 | 717 | 70 | ||
| GenBank mRNA | FJ943978.1 | - | 1 | 518 | 70 | |
More Information
Set Membership: |
|

RAB9A
Genomic Map