Assay Details
| Gene Symbol: ELAVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166194.1 | NP_001159666.1 | 3 - 4 | 430 | 78 | |
| XM_015095474.2 | XP_014950960.1 | 4 - 5 | 593 | 78 | ||
| GenBank mRNA | GQ221065.1 | - | 3 - 4 | 430 | 78 | |
More Information
Set Membership: |
|

ELAVL1
Genomic Map