Assay Details
| Gene Symbol: TRA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318073.1 | NP_001305002.1 | 7 - 8 | 777 | 68 | |
| XM_012097668.2 | XP_011953058.1 | 7 - 8 | 6055 | 68 | ||
| XM_027965566.1 | XP_027821367.1 | 8 - 9 | 1214 | 68 | ||
| XM_027965630.1 | XP_027821431.1 | 9 - 10 | 1388 | 68 | ||
| GenBank mRNA | KT186101.1 | - | 7 - 8 | 777 | 68 | |
More Information
Set Membership: |
|

TRA2B
Genomic Map