Assay Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009243.1 | NP_001009243.1 | 4 - 5 | 540 | 69 | |
| GenBank mRNA | AF290202.1 | - | 4 - 5 | 540 | 69 | |
| S80098.1 | - | 2 - 3 | 172 | 69 | ||
More Information
Set Membership: |
|

STAR
Genomic Map