Assay Details
| Gene Symbol: PPIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163048.1 | NP_001156520.1 | 9 - 10 | 839 | 57 | |
| XM_012102171.3 | XP_011957561.1 | 9 - 10 | 1301 | 57 | ||
| GenBank mRNA | FJ969405.1 | - | 9 - 10 | 839 | 57 | |
More Information
Set Membership: |
|

PPIE
Genomic Map