Assay Details
| Gene Symbol: IFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009803.1 | NP_001009803.1 | 3 - 4 | 418 | 63 | |
| GenBank mRNA | AY575609.1 | - | 1 - 2 | 50 | 63 | |
| AY575610.1 | - | 1 - 2 | 50 | 63 | ||
| DQ028767.1 | - | 3 - 4 | 294 | 63 | ||
| DQ410715.1 | - | 3 - 4 | 366 | 63 | ||
| X52640.1 | - | 418 | 63 | |||
More Information
Set Membership: |
|

IFNG
Genomic Map