Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009805.1 | NP_001009805.1 | 1 - 2 | 169 | 76 | |
| XM_012177585.3 | XP_012032975.1 | 1 - 2 | 330 | 76 | ||
| GenBank mRNA | X53561.1 | - | 169 | 76 | ||
| X55991.1 | - | 156 | 76 | |||
More Information
Set Membership: |
|

CSF2
Genomic Map