Assay Details
| Gene Symbol: EEF1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009449.1 | NP_001009449.1 | 4 - 5 | 344 | 60 | |
| XM_027972755.1 | XP_027828556.1 | 4 - 5 | 1376 | 60 | ||
| GenBank mRNA | AF544990.1 | - | 4 - 5 | 344 | 60 | |
More Information
Set Membership: |
|

EEF1D
Genomic Map