Assay Details
| Gene Symbol: CFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009484.2 | NP_001009484.1 | 1 - 2 | 322 | 75 | |
| GenBank mRNA | AY676116.1 | - | 1 - 2 | 323 | 75 | |
More Information
Set Membership: |
|

CFL1
Genomic Map