Assay Details
| Gene Symbol: CD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129902.1 | NP_001123374.1 | 8 - 9 | 1344 | 70 | |
| XM_027965769.1 | XP_027821570.1 | 9 - 10 | 1541 | 70 | ||
| GenBank mRNA | DQ407520.1 | - | 8 - 9 | 1344 | 70 | |
More Information
Set Membership: |
|

CD4
Genomic Map