Assay Details
| Gene Symbol: RAB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161861.1 | NP_001155333.1 | 3 - 4 | 604 | 92 | |
| GenBank mRNA | FJ943969.1 | - | 3 - 4 | 604 | 92 | |
More Information
Set Membership: |
|

RAB10
Genomic Map