Assay Details
| Gene Symbol: SLC7A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252182.1 | NP_001239111.1 | 7 - 8 | 877 | 57 | |
| XM_012097270.3 | XP_011952660.1 | 7 - 8 | 1549 | 57 | ||
| GenBank mRNA | JF719289.1 | - | 7 - 8 | 877 | 57 | |
| JQ085938.1 | - | 7 - 8 | 913 | 57 | ||
More Information
Set Membership: |
|

SLC7A11
Genomic Map