Assay Details
| Gene Symbol: IL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009808.1 | NP_001009808.1 | 3 - 4 | 318 | 70 | |
| XM_012159802.3 | XP_012015192.2 | 4 - 5 | 400 | 70 | ||
| XM_027965767.1 | XP_027821568.1 | 3 - 4 | 1798 | 70 | ||
| XM_027965768.1 | XP_027821569.1 | 3 - 4 | 365 | 70 | ||
| GenBank mRNA | X60167.1 | - | 3 - 4 | 318 | 70 | |
More Information
Set Membership: |
|

IL1A
Genomic Map