Assay Details
| Gene Symbol: CD80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024963.2 | NP_001020134.1 | 2 - 3 | 382 | 68 | |
| XM_012174077.3 | XP_012029467.1 | 2 - 3 | 624 | 68 | ||
| GenBank mRNA | AY390554.1 | - | 2 - 3 | 385 | 68 | |
| AY390555.1 | - | 2 - 3 | 385 | 68 | ||
More Information
Set Membership: |
|

CD80
Genomic Map