Assay Details
| Gene Symbol: SERPINE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174114.2 | NP_001167585.1 | 6 - 7 | 1137 | 59 | |
| GenBank mRNA | GQ855215.1 | - | 6 - 7 | 1140 | 59 | |
More Information
Set Membership: |
|

SERPINE1
Genomic Map